Learn More
Grant Instruments™ Platform for Grant Orbital Shakers and Shaker Incubators
Includes clamps to keep flasks and beakers from spilling. Grant Instruments™ Platform for Grant Orbital Shakers and Shaker Incubators are interchangeable to accommodate a variety of vessels.
Brand: Grant Instruments™ P6-1000
Code : 04
Additional Details : Weight : 3.50000kg
Description
- Includes clamps to keep vessels secure
- For use with
- ES-20 Shaker Incubator
- ES-80 Shaker Incubator
- PSU 10i Orbital Shaker
- PSU 20i Orbital Shaker
Specifications
6 x 1000 mL Flasks | |
PSU-20i Orbital Platform Shaker | |
6 |