Learn More
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators
Accommodates tubes of different diameters. Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators keeps test tubes secure during rotation
£68.35 - £151.00
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
11411078
|
Grant Instruments™
PRS-10 |
10 Tubes up to 30 mm, Tube volumes up to 50 mL |
£68.35
Each |
In Stock
Log in to see stock. |
||||||
11421028
|
Grant Instruments™
PRS-14 |
14 tubes up to 30mm, tube volumes up to 50mL |
£126.27
Each |
In Stock
Log in to see stock. |
||||||
11431028
|
Grant Instruments™
PRS-22 |
22 Tubes up to 15 mm, Tube volumes up to 15 mL |
£89.55
Each |
In Stock
Log in to see stock. |
||||||
11491068
|
Grant Instruments™
PRS-26 |
26 Tubes up to 15 mm, 1.5 -15 mL |
£96.30
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11411028
|
Grant Instruments™
PRS-48 |
48 Tubes up to Ø 15 mm, Tube volumes 1.5-15 mL |
£151.00
Each |
In Stock
Log in to see stock. |
||||||
10860891
|
Grant Instruments™
PRS-5-12 |
5 Tubes up to 30 mm and 12 Tubes up to 15 mm, Tube volumes 50 mL/1.5-15 mL |
£78.55
Each |
In Stock
Log in to see stock. |
||||||
12915511
|
Grant Instruments™
PRS-8-22 |
8 tubes up to 30 mm plus 22 tubes up to 15 mm, tube volumes up to 50 mL/1.5-15 mL |
£131.00
Each |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||||
Description
- For use with PTR-35 and PTR-60 Series 360° vertical multi-function rotators
- Hold as few as 8 test tubes or as many as 48.
- Accommodates a variety of capacities
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators