missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Plastic Tube Brushes

A durable, 100% polypropylene brush that cleans tubes and small parts without scratching finely polished stainless steel surfaces and is flexible enough to pass through tank elbow fittings.

Brand:  Micronova™ BRPL-25S36

67.32 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


Product Code. 15743807

  • £71.60 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Micronova Plastic Tube brushes are made from FDA compliant, 100% polypropylene bristles and handles. The narrow center cores, also polypropylene, are flexible enough to pass through tank elbow fittings.

  • Durable plastic brush for tubes and small parts
  • 100% polypropylene bristles and handle
  • Will not scratch finely polished stainless steel
  • Flexible enough to bend through fittings
  • FDA Compliant materials
Specifications

Specifications

Flexible Tube Brush
Polypropylene
Non-sterile
White
6.4 cm
Tube Brush to Clean Tubing and Small Parts
5 in.
91.4 cm
2 in.
Polypropylene
Flexible
Keep at ambient temperature
2.5 in.
Reusable
36 in.
1/4 in.
Chemicals
5.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers