missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum, 1% on polyethyleneimine/SiO2, Thermo Scientific™
Reduction reactions
£183.00
Chemical Identifiers
CAS | 7440-06-4 |
---|---|
Molecular Formula | Pt |
Molecular Weight (g/mol) | 195.08 |
MDL Number | MFCD00011179 |
InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
Synonym | black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion |
PubChem CID | 23939 |
ChEBI | CHEBI:33400 |
IUPAC Name | platinum |
SMILES | [Pt] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375687
|
Thermo Scientific Alfa Aesar
043407.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Specifications
7440-06-4 | |
Pt | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
Odorless |
Beads | |
MFCD00011179 | |
Soluble in aqua regia. Insoluble in water,mineral acids and organic acids. | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
Platinum, 1% on polyethyleneimine/SiO2 |
Safety and Handling
GHS H Statement
H332-H335
Harmful if inhaled.
May cause respiratory irritation.
P261-P285-P304+P341-P342+P311-P501c
H334
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures