missing translation for 'onlineSavingsMsg'
Learn More

Platinum 30 wt% Rhodium wire, 0.33mm (0.013in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 045571.BW

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15453995

  • POA / 50cm
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum 30 wt% Rhodium wire
Pt:Rh; 70:30 wt%
50 cm
≈0.75g/50cm
0.33mm (0.013 in.) dia., ISA Type B Standard Grade Thermocouple
MFCD00198186
Odorless
Wire
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only