missing translation for 'onlineSavingsMsg'
Learn More

Platinum 30 wt% Rhodium wire, 0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals

For ISA Type B Standard Grade Thermocouple

Brand:  Thermo Scientific Alfa Aesar 012576.BU

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11337427

  • £1985.00 / 25cm
Estimated Shipment: 05-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Platinum 30 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type B Standard Grade Thermocouple is used in the glass industry for furnace linings and complex fabrications. It is used as a catalyst in the preparation of nitric acid and in rayon spinnerets. It is used as a device for measuring high temperatures. It serves as a catalyst in the oxidation of the ammonia by atmospheric oxygen to produce nitric oxide, through Haber process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum 30 wt% Rhodium wire
Wire
MFCD00198186
Odorless
0.5mm (0.02 in.) dia.
25 cm
Insoluble in water.
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only