missing translation for 'onlineSavingsMsg'
Learn More

Platinum on activated carbon, 5% Pt, Thermo Scientific Chemicals

£116.58 - £391.95

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10318290
View Documents Promotion Details
Thermo Scientific Acros
195230100
10 g Glass bottle
£116.58
10g
Please to purchase this item. Need a web account? Register with us today!
10266730
View Documents
Thermo Scientific Acros
195230500
50 g Glass bottle
£391.95
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

Black
Pt
01,982; 08,405; 04,393; 04,393; 04,393; 04,393
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
195.08
Particle size distribution: <7 Micron: 10%, <28 Micron: 50%, <85 Micron: 90%
Platinum on activated carbon, 5% Pt
7440-06-4
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
5% (Pt)
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Platinum on activated carbon

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only