missing translation for 'onlineSavingsMsg'
Learn More

Platinum black, HiSPEC™ 1000, Thermo Scientific Chemicals

Product Code. 11367537
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Platinum black is most commonly used for electrolyte fuel cell, microbial fuel cell, polymer electrolyte fuel cell and phosphoric acid fuel cells. It is used as a catalyst in hydrogenation reactions and a gas adsorbent. It can be coated on electrode surface by electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Appearance Dark gray powder
Quantity 1 g
Solubility Information Platinum black is insoluble in water and soluble in aqua regia.
Odor Odorless
Form Powder
Chemical Name or Material Platinum black

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'