missing translation for 'onlineSavingsMsg'
Learn More

Platinum Cobalt Color Standard APHA 500), Standard, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals H296-03

646.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.



Additional Details : Weight : 1.00000kg Transport : UN number : 3264 Chem class : 8 Pack group : II


 View more versions of this product

Product Code. 15547734

  • £680.00 / 500mL
Estimated Shipment: 14-11-2024
to see stock.

Description

Description

Specifications

Specifications

Platinum Cobalt Color Standard (APHA 500)
500 mL
Solution
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B

Hazard Statement
  • H350i-May cause cancer by inhalation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P281-Use personal protective equipment as required.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers