missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical™
£70.80
Specifications
Color | Yellow |
---|---|
pH Range | 1.1 |
Format | Liquid |
Packaging | Glass Bottle |
Type | Platinum Cobalt Color Standard Solution |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11987175
|
Fisher Bioreagents
USSP120500 |
500 mL | Glass Bottle |
£70.80
500mL |
Estimated Shipment: 18-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Yellow | |
Liquid | |
Platinum Cobalt Color Standard Solution |
1.1 | |
Glass Bottle |