missing translation for 'onlineSavingsMsg'
Learn More

Platinum Crucible With Lid

Available in 10mL to 50mL capacities

Brand:  N/A 8411 2025

 View more versions of this product

Product Code. 12867673

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Quoted masses are approximate.
Approximate process temperature 1,200°C and maximum temperature is 1,400°C (for short time of 10min)
Specifications

Specifications

Platinum
1Each
35mm
Crucible
1200°C
25 mL
37mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.