missing translation for 'onlineSavingsMsg'
Learn More

Platinum foil, 0.1mm (0.004in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

£311.00 - £994.95

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011509.FF
25 x 25 mm
£311.00
25mm
Please to purchase this item. Need a web account? Register with us today!
11375879
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011509.FI
50 x 50 mm
£994.95
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

7440-06-4
21.45g/cm3 at 20°C
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Foil
Silvery white foil
99.99% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Odorless
Platinum
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-116-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only