missing translation for 'onlineSavingsMsg'
Learn More

Platinum gauze, 52 mesh woven from 0.1mm (0.004in) dia wire, 99.9% (metals basis)

Product Code. p-5545476
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
50 x 75 mm
75 x 75 mm
100 x 100 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11301368

Brand: Thermo Scientific Alfa Aesar 010283.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining. | CAS: 7440-06-4 | Pt | 195.08 g/mol

It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Quantity 25 x 25 mm
Mesh Size 52 Mesh
Packaging Shrink-wrapped
Form Wire Cloth
Solubility Information Insoluble in water. Dissolves in aqua regia
Percent Purity 99.9%
Chemical Name or Material Platinum gauze

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.