missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) 2,4-pentanedionate, Pt 48.0% min, Thermo Scientific Chemicals

Product Code. 11306048
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for photoactivated hydrosilylation reactions between vinyl and silane groups; in the deposition of platinum metal and in the preparation of platinum nanotubes. Precursor for FePt superlattice magnetic nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15170-57-7
Molecular Formula C10H14O4Pt
Molecular Weight (g/mol) 393.30
MDL Number MFCD00000028
InChI Key KLFRPGNCEJNEKU-FDGPNNRMSA-L
Synonym platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii
PubChem CID 10960186
SMILES [Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 250°C to 252°C
Color Red
Physical Form Powder
Quantity 1 g
Solubility Information Very soluble in chloroform; insoluble in water
IUPAC Name platinum(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Formula Weight 393.31
Chemical Name or Material Platinum(II) 2, 4-pentanedionate, Pt 48.0% min.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'