missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) chloride, 99.9% (metals basis), Pt 73% min, Thermo Scientific Chemicals

£210.05 - £818.07

Chemical Identifiers

CAS 10025-65-7
Molecular Formula Cl2Pt
Molecular Weight (g/mol) 265.98
MDL Number MFCD00011181
InChI Key CLSUSRZJUQMOHH-UHFFFAOYSA-L
IUPAC Name platinum(2+) dichloride
SMILES [Cl-].[Cl-].[Pt++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311688
View Documents
Thermo Scientific Alfa Aesar
011044.03
1 g
£210.05
1g
Please to purchase this item. Need a web account? Register with us today!
11301688
View Documents
Thermo Scientific Alfa Aesar
011044.06
5 g
£818.07
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II). It is a well known chemotherapy drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10025-65-7
265.98
CLSUSRZJUQMOHH-UHFFFAOYSA-L
[Cl-].[Cl-].[Pt++]
Cl2Pt
MFCD00011181
platinum(2+) dichloride

Specifications

581°C (decomposition)
Brown-Green
Cl2Pt
MFCD00011181
14,7528
CLSUSRZJUQMOHH-UHFFFAOYSA-L
platinum(2+) dichloride
266
Pt 73% min.
(metals basis)
10025-65-7
Crystalline
PtCl2
UN3261
Soluble in ammonia and hydrochloric acid; Insoluble in water,alcohol and ether.
[Cl-].[Cl-].[Pt++]
265.98
99.9%
5.87 g/mL
Platinum(II) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P260-P264b-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H334

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 233-034-1

missing translation for 'rtecsNumber' : TP2275000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Platinum(II) chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Platinous chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only