missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) chloride, Premion™, 99.99+% (metals basis), Pt 73% min, Thermo Scientific Chemicals

Catalog Number 11344748
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10025-65-7
Cl2Pt
265.98
MFCD00011181
CLSUSRZJUQMOHH-UHFFFAOYSA-L
platinum(2+) dichloride
[Cl-].[Cl-].[Pt++]
This item is not returnable. View return policy
10025-65-7
Cl2Pt
265.98
MFCD00011181
CLSUSRZJUQMOHH-UHFFFAOYSA-L
platinum(2+) dichloride
[Cl-].[Cl-].[Pt++]

Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 581°C (decomposition)
Color Brown-Green
Physical Form Powder
Quantity 5 g
Linear Formula PtCl2
UN Number UN3260
Merck Index 14,7528
Solubility Information Soluble in ammoniaband hydrochloric acid. Insoluble in water,alcohol and ether.
Formula Weight 266
Percent Purity ≥99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pt 73% min
Density 5.87 g/mL
Assay (metals basis)
Chemical Name or Material Platinum (II) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'