missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ II Hot-Start PCR Master Mixes (2X)

Invitrogen Platinum II Hot-Start PCR Master Mix (2X) offers Platinum II Taq Hot-Start DNA Polymerase premixed with Platinum II PCR buffer and dNTPs for convenient PCR setup.

Brand:  Invitrogen™ 14000012

56.06 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15889844

  • £74.75 / 50 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 1.25 mL Platinum II PCR Master Mix (2X), 1.25 mL Platinum GC Enhancer, 1.25 mL Water, nuclease-free
This item is not returnable. View return policy

Description

Description

Invitrogen Platinum II Hot-Start PCR Master Mix (2X), available in colorless and green formats, offers Platinum II Taq Hot-Start DNA Polymerase premixed with Platinum II PCR buffer and dNTPs for convenient PCR setup. Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase and leading hot-start technology. This master mix is provided with the optional Platinum GC Enhancer for specific amplification and improved yields of GC-rich targets.

Platinum II Hot-Start Green PCR Master Mix also includes two tracking dyes for direct loading of PCR products on gels.

Features

  • Innovative buffer enables universal annealing temperature by isostabilizing primer-template duplex structures
  • Engineered Taq DNA polymerase confers fast cycling and resistance to common inhibitors
  • Hot-start technology enables superior specificity, sensitivity, and yields and allows for room temperature reaction setup
  • Green PCR buffer reduces pipetting errors with direct gel loading

Applications

  • Amplification of DNA from complex genomic, viral, and plasmid templates
  • Amplification and improved yields of GC-rich targets
  • RT-PCR
  • Genotyping
  • High-throughput PCR

Platinum II Taq Hot-Start DNA Polymerase is an engineered Taq DNA polymerase that shows increased resistance to reaction inhibitors originating from sample material or DNA purification steps. The polymerase has a higher DNA synthesis rate and delivers PCR results more than two times faster than other Taq DNA polymerases. Proprietary Platinum Taq antibodies block polymerase activity at ambient temperatures and dissociate after the initial denaturation step at 94°C. This automatic hot start provides increased sensitivity, specificity, and yield, while allowing reaction assembly at room temperature.

Due to the unique composition of the Platinum II PCR buffer, the annealing temperature is 60°C for most primer pairs designed following the general design rules. Isostabilizing molecules in the buffer increase primer–template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize annealing temperature for each primer pair. With Platinum II Hot-Start PCR Master Mix (2X), different PCR assays can be cycled together using the same protocol with universal primer annealing temperature and the extension step selected for the longest fragment to be amplified.

Notes

  • For applications that require PCR product analysis by absorbance or fluorescence excitation, colorless format is recommended.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Platinum II PCR Master Mix (2X), 1.25 mL
• Platinum GC Enhancer, 1.25 mL
• Nuclease-free water, 1.25 mL

Store at -20°C in a non-frost-free freezer.
High
Fast or Standard
HPLC
Dry Ice
2X
Built-In Hot Start
3'-A
5 kb or less
Colorless
Tube
Platinum II Taq Hot-Start DNA Polymerase
Hot Start PCR Master Mix
50 reactions
Hot-start PCR
1X
50 reactions
SuperMix or Master Mix
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.