missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ II Taq Hot-Start DNA Polymerase

Invitrogen Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology.

Brand:  Invitrogen™ 14966000

3.06 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 15949082

  • £4.59 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price


Includes: 10 μL Platinum II Taq HS DNA Polymerase, 1.25 mL 5X Platinum II PCR Buffer, 1.25 mL Platinum GC Enhancer
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Taq HS DNA Polymerase
• PCR buffer
• GC enhancer

Store at -20°C.
High
Fast or Standard
Platinum™
25 Reactions
Genotyping
1X
3'-A
5 kb or less
Tube
Platinum II Taq Hot-Start DNA Polymerase
5' to 3'
Hot Start DNA Polymerase
Dry Ice
2X
25 Reactions
Separate Components
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Platinum™ II Taq Hot-Start DNA Polymerase > 25 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.