missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings.

Brand:  Thermo Scientific Alfa Aesar 041802.FI

494.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11375929

  • £496.47 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis) is used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Platinum Iridium foil
Pt:Ir; 90:10 wt%
Insoluble in water.
Tube
50 x 50 mm
MFCD02091683
99.9%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN18L048 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog Number041802.FI,041802.GH,041802.FF
Lot NumberZ09H099 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number041802.FI,041802.GH,041802.FF
Lot NumberP23C013 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041802.FI,041802.GH,041802.FF
Lot NumberU14E033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041802.FI,041802.GH,041802.FF
Lot NumberI30T017 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041802.FI,041802.GH,041802.FF
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals > 50 x 50mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only