missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium gauze, 150 mesh woven from 0.043mm (0.0017in) dia wire

Product Code. 11410351
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11410351

Brand: Thermo Scientific Alfa Aesar 040934.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Filtration, electrodes

Filtration, electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Iridium gauze
Name Note Woven from 0.043mm (0.0017 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
Quantity 50 x 50 mm
Mesh Size 150 Mesh
Odor Odorless
Form Wire Cloth
MDL Number MFCD02091683

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.