Learn More
Platinum Iridium wire, 0.127mm (0.005in) dia, annealed, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals
£45.50 - £193.00
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364658
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
043019.BS
10 cm
£45.50
10cm
View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2024
to see stock.
11374658
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
043019.BW
50 cm
£193.00
50cm
View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2024
to see stock.
Description
Description
Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes.Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes