missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Iridium wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 cm
50 cm
Description
Used in the fabrication of metal microelectrodes for electrical stimulation of nervous tissues and in electrophysiological recordings. Inoculating needles and leadwires for implants. Used in electrode materials, composing the gel boxes/electrophoresis chambers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Silver to Gray |
Appearance | Silver gray wire |
Assay Percent Range | >99.9% |
Quantity | 10 cm |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Form | Wire |
Chemical Name or Material | Platinum Iridium |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'