missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Iridium wire, 0.2mm (0.008in) dia, annealed, 99.9+% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 cm
50 cm
Description
Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Silver to Gray |
Appearance | Silver gray wire |
Assay Percent Range | >99.9% |
Quantity | 10 cm |
Solubility Information | Insoluble in water and acid. |
Odor | Odorless |
Form | Wire |
Chemical Name or Material | Platinum Iridium |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'