missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.5mm (0.02in) dia, hard, Thermo Scientific Chemicals

Form: Approx. 0.44g/10cm, Temper: as drawn

£109.55 - £2032.11

Chemical Identifiers

CAS 7440-6-6
Molecular Formula Pt:Ir; 90:10 wt%
MDL Number MFCD02091683
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336767
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010056.BS
10 cm
£109.55
10cm
Please to purchase this item. Need a web account? Register with us today!
11326767
View Documents
Thermo Scientific Alfa Aesar
010056.BW
50 cm
£409.04
50cm
Please to purchase this item. Need a web account? Register with us today!
11346767
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
010056.CB
250 cm
£2032.11
250cm
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fabrication of metal microelectrodes for electrical stimulation of nervous tissue and electrophysiological recordings. Inoculating needles and leadwires for implants. Electrode material for gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécification

Identifiants chimiques

7440-6-6
Pt:Ir; 90:10 wt%
MFCD02091683

Spécification

White
Silvery white wire
MFCD02091683
Odorless
Platinum, Hard
7440-6-6
Pt:Ir; 90:10 wt%
Insoluble in water
Wire
Vidéos
Sécurité et traitement

Sécurité et traitement

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Certificats

RUO – Research Use Only