Learn More
Platinum Iridium wire, 0.5mm (0.02in) dia, hard, Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 010055.BS
 View more versions of this product
Product Code. 11306767
  • £103.52 / 10cm
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Platinum Iridium wire
Pt:Ir; 85:15 wt%
Shrink-wrapped
10 cm
MFCD02091683