missing translation for 'onlineSavingsMsg'
Learn More
Platinum(IV) bromide, 99.99% (metals basis), Pt 37.1% min, Thermo Scientific Chemicals Product Code.: 11396548

Platinum(IV) bromide, 99.99% (metals basis), Pt 37.1% min, Thermo Scientific Chemicals

Product Code. 11396548
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11396548

Brand: Thermo Scientific Alfa Aesar 012171.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Platinum(IV) bromide can be used as an analytical reagent to test for the presence of cesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68938-92-1
Molecular Formula Br4Pt
Molecular Weight (g/mol) 514.70
MDL Number MFCD00016289
InChI Key DYFRBYUUSCNRBP-UHFFFAOYSA-J
Synonym platinum tetrabromide, platinum iv bromide, platinum 4+ tetrabromide
IUPAC Name tetrabromoplatinumtetrakis(ylium)
SMILES Br[Pt+4](Br)(Br)Br

Specifications

Melting Point 180°C (decomposition)
Color Brown to Black
Physical Form Crystalline
Quantity 5 g
Linear Formula PtBr4
UN Number UN3260
Formula Weight 514.73
Percent Purity 99.99%
Concentration or Composition (by Analyte or Components) Pt 37.1% min.
Assay (metals basis)
Chemical Name or Material Platinum(IV) bromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ18K014 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number012171.03,012171.06
Lot NumberY15J026 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number012171.03,012171.06
Lot NumberU05I023 Certificate TypeCertificate of Analysis Date12/08/2022 Catalog Number012171.03,012171.06
Lot NumberY15H098 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012171.03,012171.06
Lot NumberX18H058 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012171.03,012171.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum(IV) bromide, 99.99% (metals basis), Pt 37.1% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.