missing translation for 'onlineSavingsMsg'
Learn More
Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min, Thermo Scientific Chemicals Product Code.: 11321688

Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min, Thermo Scientific Chemicals

Product Code. 11321688
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11321688

Brand: Thermo Scientific Alfa Aesar 011045.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Platinum(IV) chloride is used as a catalyst and analytical reagent. Further, it is used in the production of semiconductor epitaxy and in electroplating process. It is involved in the preparation of platinum(II) chloride .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-96-1
Molecular Formula Cl4Pt
Molecular Weight (g/mol) 336.88
MDL Number MFCD00011182
InChI Key FBEIPJNQGITEBL-UHFFFAOYSA-J
Synonym platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4Show MoreShow Less
PubChem CID 26031
IUPAC Name tetrachloroplatinum
SMILES Cl[Pt](Cl)(Cl)Cl

Specifications

Melting Point 370°C (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water and acetone. Slightly soluble in ethanol and insoluble in ether.
Formula Weight 336.9
Sensitivity Hygroscopic
Density 4.303 g/cm3 at 20°C
Chemical Name or Material Platinum(IV) chloride, Pt 57% min.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU16K034 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog Number011045.03,011045.06
Lot NumberS21K020 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number011045.03,011045.06
Lot NumberP25K009 Certificate TypeCertificate of Analysis Date08/04/2024 Catalog Number011045.03,011045.06
Lot NumberN13K027 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number011045.03,011045.06
Lot NumberW05J024 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog Number011045.03,011045.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.