missing translation for 'onlineSavingsMsg'
Learn More
Platinum(IV) chloride, 99%, Thermo Scientific Chemicals Product Code.: 10541451

Platinum(IV) chloride, 99%, Thermo Scientific Chemicals

Product Code. 10541451
5 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10541451

Brand: Thermo Scientific Acros 195400050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-96-1
Molecular Formula Cl4Pt
Molecular Weight (g/mol) 336.88
MDL Number MFCD00011182
InChI Key FBEIPJNQGITEBL-UHFFFAOYSA-J
Synonym platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4Show MoreShow Less
PubChem CID 26031
IUPAC Name tetrachloroplatinum
SMILES Cl[Pt](Cl)(Cl)Cl

Specifications

Melting Point 370.0°C
Color Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99%
Linear Formula PtCl4
Solubility Information Solubility in water: 587g/L (25°C). Other solubilities: alcohol
Formula Weight 336.89
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Platinum(IV) chloride, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465018 Certificate TypeCertificate of Analysis Date09/08/2024 Catalog Number195402500,195400010,195400050
Lot NumberA0461879 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number195402500,195400010,195400050
Lot NumberA0457923 Certificate TypeCertificate of Analysis Date04/12/2023 Catalog Number195402500,195400010,195400050
Lot NumberA0455851 Certificate TypeCertificate of Analysis Date31/08/2023 Catalog Number195402500,195400010,195400050
Lot NumberA0451369 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number195402500,195400010,195400050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Platinum(IV) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Platinum(IV) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum(IV) chloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.