Learn More
Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min, Thermo Scientific Chemicals
£135.07 - £598.98
Chemical Identifiers
CAS | 13454-96-1 |
---|---|
Molecular Formula | Cl4Pt |
Molecular Weight (g/mol) | 336.88 |
MDL Number | MFCD00011182 |
InChI Key | FBEIPJNQGITEBL-UHFFFAOYSA-J |
Synonym | platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4 |
PubChem CID | 26031 |
IUPAC Name | tetrachloroplatinum |
SMILES | Cl[Pt](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435220
|
Thermo Scientific Alfa Aesar
044503.03 |
1 g | N/A | ||||||
11445220
|
Thermo Scientific Alfa Aesar
044503.06 |
5 g |
£598.98
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum(IV) chloride is used as a catalyst and analytical reagent. Further, it is used in the production of semiconductor epitaxy and in electroplating process. It is involved in the preparation of platinum(II) chloride .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13454-96-1 | |
336.88 | |
FBEIPJNQGITEBL-UHFFFAOYSA-J | |
26031 | |
Cl[Pt](Cl)(Cl)Cl |
Cl4Pt | |
MFCD00011182 | |
platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4 | |
tetrachloroplatinum |
Specifications
370°C (decomposition) | |
Brown | |
Cl4Pt | |
UN2923 | |
FBEIPJNQGITEBL-UHFFFAOYSA-J | |
tetrachloroplatinum | |
26031 | |
≥99.99% | |
Pt 57% min | |
Hygroscopic | |
(metals basis) |
13454-96-1 | |
Crystalline Soluble | |
MFCD00011182 | |
platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4 | |
Cl[Pt](Cl)(Cl)Cl | |
336.88 | |
336.9 | |
Premion™ | |
Odorless | |
4.303g/mL | |
Platinum (IV) chloride |
Safety and Handling
GHS H Statement
H301-H334-H314-H318-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
missing translation for 'rtecsNumber' : TP2275500
missing translation for 'tsca' : TSCA
RUO – Research Use Only