missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) oxide hydrate, typical Pt-content 79-84%, Thermo Scientific Chemicals

Product Code. 10552231
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52785-06-5
Molecular Formula O2Pt
Molecular Weight (g/mol) 227.08
MDL Number MFCD00066964
InChI Key YKIOKAURTKXMSB-UHFFFAOYSA-N
Synonym Adams' catalyst
IUPAC Name dioxoplatinum
SMILES O=[Pt]=O

Specifications

Melting Point 450.0°C
Color Brown to Black
Physical Form Powder
Quantity 1 g
Linear Formula PtO2
UN Number 1479
Fieser 01,890; 15,268
Merck Index 14, 7527
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. acids and dil.sol. of koh
Formula Weight 227.09
Percent Purity 79 to 84% Pt
Packaging Glass bottle
Chemical Name or Material Platinum(IV) oxide hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0396671 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number10629103,211920050,211920010,211920250
Lot NumberA0434393 Certificate TypeCertificate of Analysis Date17/09/2024 Catalog Number211920050,211920010,211920250
Lot NumberA0461131 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog Number211920050,211920010,211920250
Lot NumberA0459465 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog Number211920050,211920010,211920250
Lot NumberA0458274 Certificate TypeCertificate of Analysis Date30/11/2023 Catalog Number211920050,211920010,211920250
compliance-icons compliance-icons
Product Identifier
  • Platinum(IV) oxide hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum(IV) oxide hydrate, typical Pt-content 79-84%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.