missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum(IV) oxide hydrate, typical Pt-content 79-84%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
missing translation for 'unitSize'
1g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Melting Point | 450.0°C |
| Color | Brown to Black |
| Physical Form | Powder |
| Quantity | 25 g |
| Linear Formula | PtO2 |
| UN Number | 1479 |
| Fieser | 01,890; 15,268 |
| Merck Index | 14, 7527 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in conc. acids and dil.sol. of koh |
| Formula Weight | 227.09 |
| Afficher plus |
Sécurité et traitement
Identifiant produit
- Platinum(IV) oxide hydrate
- Danger
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- H319-Causes serious eye irritation.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu