missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) oxide hydrate, typical Pt-content 79-84%

Product Code. 10753211 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10753211

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 52785-06-5 | O2Pt | 227.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52785-06-5
Molecular Formula O2Pt
Molecular Weight (g/mol) 227.08
MDL Number MFCD00066964
InChI Key YKIOKAURTKXMSB-UHFFFAOYSA-N
Synonym Adams' catalyst
IUPAC Name dioxoplatinum
SMILES O=[Pt]=O

Specifications

Melting Point 450.0°C
Color Brown to Black
Physical Form Powder
Quantity 5 g
Linear Formula PtO2
UN Number 1479
Fieser 01,890; 15,268
Merck Index 14, 7527
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. acids and dil.sol. of koh
Formula Weight 227.09
Percent Purity 79 to 84% Pt
Packaging Glass bottle
Chemical Name or Material Platinum(IV) oxide hydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Platinum(IV) oxide hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.