Learn More
Platinum(IV) oxide monohydrate, Adams Catalyst, Pt 75% min, Thermo Scientific Chemicals
Description
Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groupsPlatinum(IV) oxide monohydrate is used as a catalyst in hydrogenation, hydrogenolysis, dehydrogenation, and oxidation reactions in organic synthesis. It also finds application in ceramic industry, inorganic chemistry and in chemical synthesis. It is utilized for the selective reduction of alkenes without reducing nitro group.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Hydrogenation catalyst
- Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups

Specifications
Specifications
Melting Point | 450°C |
Color | Black to Brown |
Physical Form | Powder |
Quantity | 1 g |
Linear Formula | PtO2·H2O |
UN Number | UN1479 |
Merck Index | 14,7527 |
Solubility Information | Insoluble in water,acid,aqua regiaSoluble in water and caustic potash solution; Insoluble in alcohol,acid and aqua regia. |
Formula Weight | 245.10 (227.09 Anhydrous) |
Concentration or Composition (by Analyte or Components) | Pt 75% min. |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N26L027 | Certificate of Analysis | 28/02/2025 | 012278.03,012278.06 |
M26I029 | Certificate of Analysis | 12/04/2022 | 012278.03,012278.06 |
H10Q17 | Certificate of Analysis | 07/12/2021 | 012278.03,012278.06 |
G15P32 | Certificate of Analysis | 07/12/2021 | 012278.03,012278.06 |
E14L38 | Certificate of Analysis | 07/12/2021 | 012278.03,012278.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.