missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) sulfide, Pt 74.8% min, Thermo Scientific Chemicals

Product Code. 11341509
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ultra high purity and high purity compositions of platinum sulfide improve both optical quality and usefulness as scientific standards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12038-21-0
Molecular Formula PtS2
Molecular Weight (g/mol) 259.20
MDL Number MFCD00016293
InChI Key RKNFJIIYAUSTJA-UHFFFAOYSA-N
Synonym platinum iv sulfide, platinum sulfide, disulfanylideneplatinum, bis sulfanylidene platinum, dithioxoplatinum, platinum disulphide, acmc-20ajvg, platinum iv sulfide 1g, platinic sulfide,, platinum iv sulfide, pt
PubChem CID 82862
IUPAC Name bis(sulfanylidene)platinum
SMILES S=[Pt]=S

Specifications

Melting Point 225°C to 250°C (decomposition)
Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 259.22
Concentration or Composition (by Analyte or Components) Pt 74.8% min.
Chemical Name or Material Platinum(IV) sulfide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'