missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum(IV) sulfide, Pt 74.8% min, Thermo Scientific Chemicals
£224.00 - £897.00
Chemical Identifiers
CAS | 12038-21-0 |
---|---|
Molecular Formula | PtS2 |
Molecular Weight (g/mol) | 259.20 |
MDL Number | MFCD00016293 |
InChI Key | RKNFJIIYAUSTJA-UHFFFAOYSA-N |
Synonym | platinum iv sulfide, platinum sulfide, disulfanylideneplatinum, bis sulfanylidene platinum, dithioxoplatinum, platinum disulphide, acmc-20ajvg, platinum iv sulfide 1g, platinic sulfide,, platinum iv sulfide, pt |
PubChem CID | 82862 |
IUPAC Name | bis(sulfanylidene)platinum |
SMILES | S=[Pt]=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351509
|
Thermo Scientific Alfa Aesar
013992.03 |
1 g |
£224.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341509
|
Thermo Scientific Alfa Aesar
013992.06 |
5 g |
£897.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ultra high purity and high purity compositions of platinum sulfide improve both optical quality and usefulness as scientific standards.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12038-21-0 | |
259.20 | |
RKNFJIIYAUSTJA-UHFFFAOYSA-N | |
82862 | |
S=[Pt]=S |
PtS2 | |
MFCD00016293 | |
platinum iv sulfide, platinum sulfide, disulfanylideneplatinum, bis sulfanylidene platinum, dithioxoplatinum, platinum disulphide, acmc-20ajvg, platinum iv sulfide 1g, platinic sulfide,, platinum iv sulfide, pt | |
bis(sulfanylidene)platinum |
Specifications
225°C to 250°C (decomposition) | |
Powder | |
MFCD00016293 | |
Insoluble in water. | |
S=[Pt]=S | |
259.20 | |
259.22 | |
Platinum(IV) sulfide |
12038-21-0 | |
PtS2 | |
platinum iv sulfide, platinum sulfide, disulfanylideneplatinum, bis sulfanylidene platinum, dithioxoplatinum, platinum disulphide, acmc-20ajvg, platinum iv sulfide 1g, platinic sulfide,, platinum iv sulfide, pt | |
RKNFJIIYAUSTJA-UHFFFAOYSA-N | |
bis(sulfanylidene)platinum | |
82862 | |
Pt 74.8% min. |
Safety and Handling
missing translation for 'einecsNumber' : 234-876-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only