missing translation for 'onlineSavingsMsg'
Learn More
Platinum Paste, Thermo Scientific Chemicals Product Code.: 11324437

Platinum Paste, Thermo Scientific Chemicals

Product Code. 11324437
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11324437

Brand: Thermo Scientific Alfa Aesar 038316.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: 900 - 1000m2 Standard, reduced, nominally 50% water wet

Platinum is used in catalytic converters, laboratory equipment, electrical application and electrodes, platinum resistance thermometers, dentistry equipment, and jewellery. It is also used in glass fiber applications, chemical processing, off-road vehicles and fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-6-6
MDL Number MFCD00011179

Specifications

Color Black
Quantity 25 g
Solubility Information Soluble in aqua regia. Insoluble in water and acid.
Odor Odorless
Appearance Black powder
Chemical Name or Material Platinum, 3% on activated carbon powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25I052 Certificate TypeCertificate of Analysis Date09/11/2022 Catalog Number038312.14,038316.14
Lot NumberU05I017 Certificate TypeCertificate of Analysis Date12/08/2022 Catalog Number038316.06,038316.14
Lot NumberR23I047 Certificate TypeCertificate of Analysis Date06/06/2022 Catalog Number038316.06,038316.14
Lot NumberA27W017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number038312.14,038316.14
Lot NumberF29K10 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number038312.14,038316.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum Paste, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.