missing translation for 'onlineSavingsMsg'
Learn More
Platinum powder, -325 mesh, 99.9+% (metals basis), Thermo Scientific Chemicals Product Code.: 11338827

Platinum powder, -325 mesh, 99.9+% (metals basis), Thermo Scientific Chemicals

Product Code. 11338827
5 g
Click to view available options
Quantity:
0.1 g
1 g
5 g
This item is not returnable. View return policy

Product Code. 11338827

Brand: Thermo Scientific Alfa Aesar 038374.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Physical Form Powder
Quantity 5 g
Percent Purity ≥99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Platinum powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN06L017 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog Number038374.03,038374.77,038374.06
Lot NumberZ18K015 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number038374.03,038374.77,038374.06
Lot NumberY05K027 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number038374.03,038374.77,038374.06
Lot NumberS17I006 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number038374.03,038374.77,038374.06,038494.18
Lot NumberS17I009 Certificate TypeCertificate of Analysis Date23/06/2022 Catalog Number038374.03,038374.77,038374.06,038494.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum powder, -325 mesh, 99.9+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.