missing translation for 'onlineSavingsMsg'
Learn More

Platinum powder, amorphous, APS <3 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Platinum powder, 99.9%, Pt, CAS Number-7440-06-4, platinum, metal, black, metal, platino, platine, iv ion, sponge, platinum, elemental, platin german, platin, 5g, (metals basis), CHEBI:33400, Hazard Class: 4.1; Packaging Group: III, 231-116-1, H228, P210-P240-P241-P280-P370+P378q

£189.00 - £626.12

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11304237
View Documents
Thermo Scientific Alfa Aesar
036704.03
1 g
£189.00
1g
Please to purchase this item. Need a web account? Register with us today!
11314237
View Documents
Thermo Scientific Alfa Aesar
036704.06
5 g
£626.12
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

7440-06-4
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Odorless
Platinum powder
Amorphous Powder
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
99.9%
(metals basis)
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III

missing translation for 'einecsNumber' : 231-116-1

missing translation for 'rtecsNumber' : TP2160000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only