missing translation for 'onlineSavingsMsg'
Learn More

Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

£228.00 - £2705.00

Specifications

Chemical Name or Material Platinum Rhodium gauze
Name Note Woven from 0.076mm (0.003 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 90:10 wt%
Assay Percent Notes (metals basis)
MDL Number MFCD00198186
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366628
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012217.FF
25 x 25 mm
£228.00
Each
Please to purchase this item. Need a web account? Register with us today!
11385919
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012217.FI
50 x 50 mm
£847.00
Each
Please to purchase this item. Need a web account? Register with us today!
11376628
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012217.GH
100 x 100 mm
£2705.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum-rhodium alloy gauze serves as a catalyst in the oxidation of the ammonia from the Haber Process by atmospheric oxygen to produce nitric oxide and subsequently nitric acid, which is used for producing nitrogen fertilizers, ammonium nitrate, adipic acid, and toluene diisocyanate (precursor for polyurethane). It has high degree of chemical inertness and stability at very high temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum Rhodium gauze
Pt:Rh; 90:10 wt%
MFCD00198186
99.9%
Odorless
Woven from 0.076mm (0.003 in.) dia. wire
(metals basis)
Insoluble in water.
80 Mesh
≈0.39g/25x25mm
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only