missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Rhodium Palladium gauze, 80 mesh knitted from 0.076mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
£215.00 - £2552.00
Specifications
Chemical Name or Material | Platinum Rhodium Palladium gauze |
---|---|
Name Note | Woven from 0.076mm (0.003 in.) dia. wire |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Rh:Pd; 90:5:5 wt% |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD00011179 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356628
|
Thermo Scientific Alfa Aesar
012216.FF |
25 x 25 mm | N/A | ||||||
11375919
|
Thermo Scientific Alfa Aesar
012216.FI |
50 x 50 mm | N/A | ||||||
11346628
|
Thermo Scientific Alfa Aesar
012216.GH |
100 x 100 mm | N/A | ||||||
Description
Platinum Rhodium Palladium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis) is used as a catalyst in the preparation of nitric acid from ammonia. It is also used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Rhodium Palladium gauze | |
Pt:Rh:Pd; 90:5:5 wt% | |
MFCD00011179 | |
99.9% | |
≈0.39g/25x25mm |
Woven from 0.076mm (0.003 in.) dia. wire | |
(metals basis) | |
Insoluble in water. | |
80 Mesh |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only