missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ SuperFi™ DNA Polymerase

Invitrogen Platinum SuperFi DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology for the highest success in PCR.

Brand:  Invitrogen™ 12351010

160.50 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15525033

  • £214.00 / 100 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen Platinum SuperFi DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology for the highest success in PCR. Featuring >300X Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.

Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.

Benefits of Platinum SuperFi DNA Polymerase

  • Exceptional >300X Taq fidelity
  • High specificity and increased yields with Platinum hot-start technology
  • Robust amplification of difficult targets including those of sub-optimal purity or high GC content
  • Room temperature reaction setup and 24 hour bench top stability of pre-assembled reactions
  • Amplification of samples with suboptimal purity

Applications

  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates
  • Template generation for sequencing
  • High-throughput PCR
  • Long PCR

Notes

  • Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Platinum SuperFi DNA Polymerase (50 μL at 2 U/μL)
• 5X SuperFi Buffer (2 x 1.25 mL)
• 5X SuperFi GC Enhancer (1.25 mL)

Store at –5°C to –30°C.
Platinum SuperFi DNA Polymerase
100 units
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Blunt
20 kb or less
High
Fast
Wet or Dry Ice
300X
100 Reactions
Separate Components
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.