Learn More
Invitrogen™ Platinum™ SuperFi™ DNA Polymerase
Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum™ hot-start technology for the highest success in PCR.
£214.00 - £3960.00
Specifications
GC-Rich PCR Performance | High |
---|---|
Polymerase | Platinum SuperFi DNA Polymerase |
Reaction Speed | Fast |
Shipping Condition | Wet or Dry Ice |
For Use With (Application) | Hot-start PCR, High-fidelity PCR |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15525033
|
Invitrogen™
12351010 |
100 Reactions |
£214.00
100 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15535033
|
Invitrogen™
12351050 |
500 Reactions |
£952.00
500 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15545033
|
Invitrogen™
12351250 |
5 x 500 Reactions |
£3960.00
2500 units |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum™ hot-start technology for the highest success in PCR. Featuring >300X Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.
Benefits of Platinum SuperFi DNA Polymerase include:
• Exceptional >300X Taq fidelity
• High specificity and increased yields with Platinum hot-start technology
• Robust amplification of difficult-to-amplify targets including those of sub-optimal purity or with ˃65% GC content
• Convenient workflow with room temperature reaction setup and 24 hr. bench top stability of pre-assembled reactions
Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.
Applications of Platinum SuperFi DNA Polymerase
• High-fidelity PCR
• Cloning and sub-cloning
• Site-directed mutagenesis
• Amplification of GC-rich templates
• Template generation for sequencing
• High-throughput PCR
• Amplification of samples with suboptimal purity
• Long PCR
• Fast PCR
Using Platinum SuperFi DNA Polymerase
Annealing rules for Platinum SuperFi DNA Polymerase are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use the Tm calculator on our website: www.thermofisher.com/tmcalculator.
Platinum SuperFi DNA Polymerase is supplied with a separate vial of SuperFi™ GC Enhancer designed for difficult and GC-rich templates (˃65% GC).
Platinum hot-start technology efficiently inhibits DNA polymerase activity at ambient temperatures, allowing convenient reaction setup at room temperature. In addition, pre-assembled PCR reactions may be stored at room temperature for up to 24 hr. prior to the PCR.
Specifications
High | |
Fast | |
Hot-start PCR, High-fidelity PCR | |
Built-In Hot Start | |
Separate Components |
Platinum SuperFi DNA Polymerase | |
Wet or Dry Ice | |
300X | |
Blunt | |
20 kb or less |
For Research Use Only. Not for use in diagnostic procedures.