missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, 0.01mm (0.0004in) dia, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15466585
Click to view available options
Quantity:
1 m
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-06-4
Molecular Formula Pt
MDL Number MFCD00011179
Quantity 1 m
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES [Pt]
IUPAC Name platinum
Molecular Weight (g/mol) 195.08
PubChem CID 23939
ChEBI CHEBI:33400
Percent Purity 99.9%
Odor Odorless
Weight ≈1.68mg/m
Form Wire
Assay (metals basis)
Chemical Name or Material Platinum wire, 0.01mm (0.0004 in.) dia., Hard
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'