missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum wire, 0.1mm (0.004in) dia, Premion™, 99.997% (metals basis)
Click to view available options
Quantity:
1 m
5 m
Unit Size:
1m
5m
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Color | White |
| Appearance | Silvery white wire |
| Density | 21.45g/cm3 at 20°C |
| Quantity | 1 m |
| Odor | Odorless |
| Assay Percent Range | 99.997% (Metals basis) |
| Form | Wire |
| Chemical Name or Material | Platinum |
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto