missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis)
Click to view available options
Quantity:
10 cm
50 cm
250 cm
Unit Size:
10cm
250cm
50cm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Appearance | Silvery white wire |
| Density | 21.45g/cm3 at 20°C |
| Quantity | 10 cm |
| Odor | Odorless |
| Assay Percent Range | 99.997% (Metals basis) |
| Form | Wire |
| Chemical Name or Material | Platinum |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu