missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEE GEE Plato Hydrometer with Thermometer
The Plato Hydrometer with Thermometer is calibrated to show the concentration of extract as a percentage by weight in brewery wort.
Brand: VEE GEE 6614TS-5
This item is not returnable.
View return policy
Description
- Calibration for use at 20°C/68°F
- Thermometer filled with SafetyBLUE liquid
Specifications
Plato Hydrometer | |
36 cm | |
Glass | |
0°C to 50°C |
0.1° | |
0 to 8.5° Plato | |
32°F to 122°F | |
20°C (68°F) |