Learn More
Kimberly-Clark™ Kimtech™ Procedure Masks
Non-sterile pleat-style masks with high bacterial and particle filtration, and certified to EN14683 Type II. The latex- and silicone-free masks reduce contamination while ensuring comfort, with a low-lint outer layer and adjustable nose seal.
Specifications
Size Standard
Sterility Non-sterile
Style Pleated
Certifications/Compliance ISO 14001, ISO 9001; Industrial Use Only
Latex Free Yes
View More Specs
Description
Description
  • Pleat style masks with earloops
  • Excellent particle and bacterial filtration efficiency to ensure the wearer’s security.
  • Certified according to EN14683 Type II, and provided with supporting technical information to demonstrate regulatory compliance.
  • Constructed from a low-lint polyethylene film outer layer for enhanced protection.
  • Contain no natural rubber latex or silicone, reducing the risks of skin irritation for the wearer.
  • The masks are provided in a single universal size.
Specifications
Specifications
Standard
Pleated
Yes
Non-sterile
ISO 14001, ISO 9001; Industrial Use Only