Learn More
BRAND™ PLTconnect™ Software
Software for PLT unit
Brand: BRAND™ 703980
125.55 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- The PLTconnect™ software gives you the additional security of knowing all your test results are documented at all times
- PLTconnect™ transmits test results for each pipette automatically and securely through the USB port on the PC, storing them there in a database or as a test certificate
- This means you always have access to test results for your pipettes, and can print out certificates as needed
Specifications
Automatically and securely transmit test results for each pipette | |
Documenting Test Results | |
PC USB port and Pipettes |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
BRAND™ PLTconnect™ Software > Description: Automatically and securely transmit test results for each pipette
Spot an opportunity for improvement?Share a Content Correction