missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLTP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand: Bio-Techne NB400-106
This item is not returnable.
View return policy
Description
PLTP Polyclonal specifically detects PLTP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PLTP | |
Polyclonal | |
Western Blot 1:500, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:200 | |
P55058 | |
PLTP | |
A partial peptide of human PLTP. [UniProt# P55058] | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse | |
Antisera |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antisera with 0.02% Sodium Azide | |
HDLCQ9, Lipid transfer protein II, phospholipid transfer protein | |
Rabbit | |
55 kDa | |
0.2 mL | |
Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction | |
5360 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction