Learn More
Memmert™ Plug-in Tube Extensions for Oven
Use these tube extensions with Memmert ovens. Memmert™ Plug-in Tube Extensions for Ovens are designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ B29718
 View more versions of this product
Product Code. 15125543
  • £40.35 / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm)
  • Angled
  • Only models U, I, S
Specifications
Specifications
Models U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 1060
Plug-in Tube Extension
57 (Inside) mm, 60.3 (Outside) mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Plug-in Tube Extensions for Oven > Plug-in tube extension; For use with Heating oven; Straight